missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzotrifluoride 98.0+%, TCI America™

Catalog No. M12155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12155G 5 g
M12151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12155G Supplier TCI America Supplier No. M12155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13630-19-8
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.14
MDL Number MFCD00153204
InChI Key DVFVNJHIVAPTMS-UHFFFAOYSA-N
Synonym 2-methylbenzotrifluoride, 1-methyl-2-trifluoromethyl benzene, 2-trifluoromethyl toluene, 1-methyl-2-trifluoromethyl-benzene, alpha,alpha,alpha-trifluoro-o-xylene, benzene, methyl trifluoromethyl, benzene, 1-methyl-2-trifluoromethyl, 2-methyl-1-trifluoromethyl benzene, trifluoromethyltoluene, pubchem1657
PubChem CID 2775324
IUPAC Name 1-methyl-2-(trifluoromethyl)benzene
SMILES CC1=CC=CC=C1C(F)(F)F

Specifications

Color Colorless
Boiling Point 126°C
Quantity 5 g
UN Number 1993
Formula Weight 160.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbenzotrifluoride