missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzotrifluoride, 97%

Catalog No. AAB2345303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2345303 1 g
AAB2345306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2345303 Supplier Thermo Scientific Chemicals Supplier No. B2345303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13630-19-8 | C8H7F3 | 160.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13630-19-8
Molecular Formula C8H7F3
Molecular Weight (g/mol) 160.14
MDL Number MFCD00153204
InChI Key DVFVNJHIVAPTMS-UHFFFAOYSA-N
Synonym 2-methylbenzotrifluoride, 1-methyl-2-trifluoromethyl benzene, 2-trifluoromethyl toluene, 1-methyl-2-trifluoromethyl-benzene, alpha,alpha,alpha-trifluoro-o-xylene, benzene, methyl trifluoromethyl, benzene, 1-methyl-2-trifluoromethyl, 2-methyl-1-trifluoromethyl benzene, trifluoromethyltoluene, pubchem1657
PubChem CID 2775324
IUPAC Name 1-methyl-2-(trifluoromethyl)benzene
SMILES CC1=CC=CC=C1C(F)(F)F

Specifications

Density 1.174
Boiling Point 125°C to 126°C
Refractive Index 1.435
Quantity 1 g
UN Number UN1993
Beilstein 1940501
Formula Weight 160.14
Percent Purity 97%
Chemical Name or Material 2-Methylbenzotrifluoride
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only