missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbenzothiazole 98.0+%, TCI America™
$244.49 - $2928.18
Chemical Identifiers
| CAS | 120-75-2 |
|---|---|
| Molecular Formula | C8H7NS |
| Molecular Weight (g/mol) | 149.211 |
| MDL Number | MFCD00005794 |
| InChI Key | DXYYSGDWQCSKKO-UHFFFAOYSA-N |
| Synonym | 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w |
| PubChem CID | 8446 |
| IUPAC Name | 2-methyl-1,3-benzothiazole |
| SMILES | CC1=NC2=CC=CC=C2S1 |
Chemical Identifiers
| 120-75-2 | |
| 149.211 | |
| DXYYSGDWQCSKKO-UHFFFAOYSA-N | |
| 8446 | |
| CC1=NC2=CC=CC=C2S1 |
| C8H7NS | |
| MFCD00005794 | |
| 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w | |
| 2-methyl-1,3-benzothiazole |
Specifications
| 120-75-2 | |
| 238°C | |
| MFCD00005794 | |
| 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w | |
| CC1=NC2=CC=CC=C2S1 | |
| 149.211 | |
| 149.21 | |
| Liquid |
| Yellow | |
| C8H7NS | |
| 25 g | |
| DXYYSGDWQCSKKO-UHFFFAOYSA-N | |
| 2-methyl-1,3-benzothiazole | |
| 8446 | |
| ≥98.0% (GC) | |
| 2-Methylbenzothiazole |
Safety and Handling
EINECSNumber : (5)-0228
RTECSNumber : DL5600000
TSCA : Yes