accessibility menu, dialog, popup

UserName

H3CSN

CAS RN 120-75-2

IUPAC Name: 2-methyl-1,3-benzothiazole
Synonyms: 2-methylbenzothiazole
Molecular Weight (g/mol): 149.21
Molecular Formula: C8H7NS
InChi Key: DXYYSGDWQCSKKO-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2S1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)