missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazole 98.0+%, TCI America™

Catalog No. M0516500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0516500G 500 g
M051625G 25 g
M0516100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0516500G Supplier TCI America Supplier No. M0516500G

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00005794
InChI Key DXYYSGDWQCSKKO-UHFFFAOYSA-N
Synonym 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
PubChem CID 8446
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES CC1=NC2=CC=CC=C2S1

Specifications

Color Yellow
Boiling Point 238°C
Quantity 500 g
Formula Weight 149.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methylbenzothiazole
EINECSNumber (5)-0228
RTECSNumber DL5600000
TSCA Yes