missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazole, 99%

CAS: 120-75-2 | C8H7NS | 149.21 g/mol

$448.88 - $448.88

Chemical Identifiers

CAS 120-75-2
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.21
InChI Key DXYYSGDWQCSKKO-UHFFFAOYSA-N
Synonym 2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
PubChem CID 8446
IUPAC Name 2-methyl-1,3-benzothiazole
SMILES CC1=NC2=CC=CC=C2S1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150381000
SDS View Documents
Thermo Scientific Chemicals
150381000
100 g Glass bottle
Each for $448.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

120-75-2
149.21
2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
2-methyl-1,3-benzothiazole
C8H7NS
DXYYSGDWQCSKKO-UHFFFAOYSA-N
8446
CC1=NC2=CC=CC=C2S1

Specifications

120-75-2
99.0
Amber to Colorless
238.0°C
Authentic
Glass bottle
1.6150 to 1.6180
27,46
2-methylbenzothiazole, benzothiazole, 2-methyl, 2-methybenzothiazole, 2-methyl-benzothiazole, benzothiazole, methyl, usaf ek-1853, unii-7lx1xe6h8w, 2-methylbenzo d thiazole, 2-methyl benzothiazole, 7lx1xe6h8w
DXYYSGDWQCSKKO-UHFFFAOYSA-N
2-methyl-1,3-benzothiazole
8446
99%
2-Methylbenzothiazole, 99%
98.5
12.0°C to 14.0°C
1.2000g/mL
102°C
98.5% min. (GC)
C8H7NS
100 g
1.2
Solubility in water: insoluble. Other solubilities: ssluble in ether and chloroform
CC1=NC2=CC=CC=C2S1
149.21
149.21
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 204-423-3

RTECSNumber : DL5600000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only