missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzothiazol-5-ol, 97%

Catalog No. aah3293709
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3293709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3293709 Supplier Thermo Scientific Chemicals Supplier No. H3293709
Only null left

CAS: 68867-14-1 | C8H7NOS | 165.21 g/mol

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 68867-14-1
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00192276
InChI Key LAKVUPMDDFICNR-UHFFFAOYSA-N
PubChem CID 699799
IUPAC Name 2-methyl-1,3-benzothiazol-5-ol
SMILES CC1=NC2=CC(O)=CC=C2S1

Specifications

Melting Point 185°C to 187°C
Quantity 10 g
Synonym 2-methylbenzo d thiazol-5-ol, 5-benzothiazolol, 2-methyl, 2-methyl-5-benzothiazolol, 2-methylbenzothiazol-5-ol, 5-benzothiazolol,2-methyl, 5-hydroxy-2-methylbenzothiazole, 2-methyl-benzothiazol-5-ol, 2-methyl-5-hydroxybenzothiazole
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Methylbenzothiazol-5-ol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only