Learn More
2-Methylbenzophenone, 98+%
CAS: 131-58-8 | C14H12O | 196.249 g/mol
Supplier: Thermo Scientific Chemicals A1477918
Description
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.
Specifications
| 2-Methylbenzophenone | |
| -18°C | |
| 309°C to 311°C | |
| C14H12O | |
| MFCD00008518 | |
| 2045469 | |
| 2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone | |
| CKGKXGQVRVAKEA-UHFFFAOYSA-N | |
| (2-methylphenyl)-phenylmethanone | |
| 67230 | |
| ≥98% |
| 131-58-8 | |
| 1.085 | |
| >110°C (230°F) | |
| 1.595 | |
| 50 g | |
| 14,7317 | |
| Insoluble in water. | |
| CC1=CC=CC=C1C(=O)C2=CC=CC=C2 | |
| 196.249 | |
| 196.25 |
Chemical Identifiers
| 131-58-8 | |
| 196.249 | |
| CKGKXGQVRVAKEA-UHFFFAOYSA-N | |
| 67230 | |
| CC1=CC=CC=C1C(=O)C2=CC=CC=C2 |
| C14H12O | |
| MFCD00008518 | |
| 2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone | |
| (2-methylphenyl)-phenylmethanone |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 205-032-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only