missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzophenone, 98+%

Catalog No. AAA1477909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1477909 10 g
AAA1477918 50 g
AAA1477930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1477909 Supplier Thermo Scientific Chemicals Supplier No. A1477909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131-58-8 | C14H12O | 196.249 g/mol

2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylbenzophenone, is used as an important raw material and intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperature and pressure.

Chemical Identifiers

CAS 131-58-8
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008518
InChI Key CKGKXGQVRVAKEA-UHFFFAOYSA-N
Synonym 2-methylbenzophenone, o-methylbenzophenone, methanone, 2-methylphenyl phenyl, phenyl o-tolyl ketone, 2-methylphenyl phenyl methanone, benzophenone, 2-methyl, 2-methylphenyl-phenylmethanone, unii-lsn1480nff, phenyl tolyl ketone, phenyl 2-tolyl ketone
PubChem CID 67230
IUPAC Name (2-methylphenyl)-phenylmethanone
SMILES CC1=CC=CC=C1C(=O)C2=CC=CC=C2

Specifications

Melting Point -18°C
Density 1.085
Boiling Point 309°C to 311°C
Flash Point >110°C (230°F)
Refractive Index 1.595
Quantity 10 g
Beilstein 2045469
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material 2-Methylbenzophenone
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-032-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only