missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)butane hydrochloride, 94%

Catalog No. p-7030020
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2754703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2754703 Supplier Thermo Scientific Chemicals Supplier No. H2754703

CAS: 75098-40-7 | C5H14ClN | 123.624 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in hydrochloric acid.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. 

Chemical Identifiers

CAS 75098-40-7
Molecular Formula C5H14ClN
Molecular Weight (g/mol) 123.624
MDL Number MFCD07777098
InChI Key ICBNZSHUWSBMCL-UHFFFAOYSA-N
Synonym n-methyl-sec-butylamine hydrochloride, 2-methylamino butane hydrochloride, methyl sec-butyl amine hydrochloride, butan-2-yl methyl amine hydrochloride, n-methyl-2-butanamine hydrochloride, n-methylbutan-2-amine hydrochloride, n-methyl-2-butylamine hydrochloride, n-methylbutan-2-amine-hydrogen chloride 1/1
PubChem CID 21273362
IUPAC Name N-methylbutan-2-amine;hydrochloride
SMILES CCC(C)NC.Cl

Specifications

Melting Point 76°C to 82°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in hydrochloric acid.
Formula Weight 123.62
Percent Purity 94%
Chemical Name or Material 2-(Methylamino)butane hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only