CAS RN 75098-40-7
IUPAC Name:
(butan-2-yl)(methyl)amine hydrochloride
Synonyms:
methyl(sec-butyl)amine hydrochloride
Molecular Weight (g/mol):
123.62
Molecular Formula:
C5H14ClN
InChi Key:
ICBNZSHUWSBMCL-UHFFFAOYNA-N
SMILES:
Cl.CCC(C)NC
1
–
1
of
1
results
1
Promotions Available
2-(Methylamino)butane hydrochloride, 94%
2-(Methylamino)butane hydrochloride, 94%
CAS: 75098-40-7 Molecular Formula: C5H14ClN Molecular Weight (g/mol): 123.624 MDL Number: MFCD07777098 InChI Key: ICBNZSHUWSBMCL-UHFFFAOYSA-N Synonym: n-methyl-sec-butylamine hydrochloride,2-methylamino butane hydrochloride,methyl sec-butyl amine hydrochloride,butan-2-yl methyl amine hydrochloride,n-methyl-2-butanamine hydrochloride,n-methylbutan-2-amine hydrochloride,n-methyl-2-butylamine hydrochloride,n-methylbutan-2-amine-hydrogen chloride 1/1 PubChem CID: 21273362 IUPAC Name: N-methylbutan-2-amine;hydrochloride SMILES: CCC(C)NC.Cl