missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetophenone, 98%

CAS: 577-16-2 | C9H10O | 134.178 g/mol

$60.38 - $550.78

Chemical Identifiers

CAS 577-16-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00008734
InChI Key YXWWHNCQZBVZPV-UHFFFAOYSA-N
Synonym 2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
PubChem CID 11340
IUPAC Name 1-(2-methylphenyl)ethanone
SMILES CC1=CC=CC=C1C(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1169506
View Documents
Thermo Scientific Chemicals
A1169506
5 g
Each for $60.38
Only null left
 
AAA1169514
View Documents
Thermo Scientific Chemicals
A1169514
25 g
Each for $176.53
Only null left
 
AAA1169522
View Documents
Thermo Scientific Chemicals
A1169522
100 g
Each for $550.78
Only null left
 
Description

Description

2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

577-16-2
134.178
YXWWHNCQZBVZPV-UHFFFAOYSA-N
11340
CC1=CC=CC=C1C(=O)C
C9H10O
MFCD00008734
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
1-(2-methylphenyl)ethanone

Specifications

577-16-2
214°C
C9H10O
MFCD00008734
907005
Soluble in ethanol. Insoluble in water.
CC1=CC=CC=C1C(=O)C
134.178
134.18
2'-Methylacetophenone
1.02
75°C (167°F)
1.5318
5 g
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
YXWWHNCQZBVZPV-UHFFFAOYSA-N
1-(2-methylphenyl)ethanone
11340
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c

H227-H312-H332

EINECSNumber : 209-408-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only