missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylacetophenone, 98%

CAS: 577-16-2 | C9H10O | 134.178 g/mol

Supplier:  Thermo Scientific Chemicals A1169514

 View more versions of this product

Catalog No. AAA1169514


Only null left
Explore available promotions

Description

Description

2'-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Methylacetophenone is possiblely used in Vanilla, cherry, almond, coumarin and various nut nuances.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2'-Methylacetophenone
1.02
75°C (167°F)
1.5318
25 g
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
YXWWHNCQZBVZPV-UHFFFAOYSA-N
1-(2-methylphenyl)ethanone
11340
98%
577-16-2
214°C
C9H10O
MFCD00008734
907005
Soluble in ethanol. Insoluble in water.
CC1=CC=CC=C1C(=O)C
134.178
134.18

Chemical Identifiers

577-16-2
134.178
YXWWHNCQZBVZPV-UHFFFAOYSA-N
11340
CC1=CC=CC=C1C(=O)C
C9H10O
MFCD00008734
2'-methylacetophenone, 1-o-tolyl ethanone, 2-methylacetophenone, o-methylacetophenone, 2-acetyltoluene, o-acetyltoluene, 1-2-methylphenyl ethanone, ethanone, 1-2-methylphenyl, 2'-methylacetylphenone, acetophenone, 2'-methyl
1-(2-methylphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P261-P271-P280-P302+P352-P304+P340-P312-P363-P370+P378q-P501c

H227-H312-H332

EINECSNumber : 209-408-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only