missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide, 95%, Thermo Scientific Chemicals

Catalog No. p-7038743
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57776ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH57776ME Supplier Thermo Scientific Chemicals Supplier No. H57776ME

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (1.00E+06 mg/L).

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.

Chemical Identifiers

CAS 1158098-73-7
Molecular Formula C7H13NO2S
Molecular Weight (g/mol) 175.246
MDL Number MFCD14702522
InChI Key VKUZMNXQGKBLHN-UHFFFAOYSA-N
Synonym 2-methyl-n-oxetan-3-ylidene propane-2-sulfinamide, 2-methyl-n-3-oxetanylidene-2-propanesulfinamide, 2-methyl-n-3-oxetanylidene propane-2-sulfinamide, 3-tert-butylsulfinylmethylene oxetane, n-tert-butylsulfinyl oxetane-3-imine, 2-methyl-n-oxetan-3-ylidene-propane-2-sulfinamide, 2-methyl-n-oxetan-3-ylidene ;propane-2-sulfinamide, 2-methyl-propane-2-sulfinic acid oxetan-3-ylideneamide, 2-methylpropane-2-sulfinic acid oxetan-3-ylideneamide
PubChem CID 46839995
IUPAC Name 2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
SMILES CC(C)(C)S(=O)N=C1COC1

Specifications

Quantity 500 mg
Solubility Information Soluble in water. (1.00E+06mg/L).
Formula Weight 175.25
Percent Purity 95%
Chemical Name or Material 2-Methyl-N-(3-oxetanylidene)propane-2-sulfinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only