accessibility menu, dialog, popup

UserName

CAS RN 1158098-73-7

IUPAC Name: 2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
Synonyms: 2-methyl-N-(oxetan-3-ylidene)propane-2-sulfinamide
Molecular Weight (g/mol): 175.25
Molecular Formula: C7H13NO2S
InChi Key: VKUZMNXQGKBLHN-UHFFFAOYNA-N
SMILES: CC(C)(C)S(=O)N=C1COC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results