missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-8-quinolinol 98.0+%, TCI America™
Supplier: TCI America M0420100G
Specifications
| 2-Methyl-8-quinolinol | |
| 73°C | |
| C10H9NO | |
| 100 g | |
| 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol | |
| CC1=NC2=C(C=CC=C2O)C=C1 | |
| 159.188 | |
| 159.19 | |
| Crystalline Powder |
| 826-81-3 | |
| 145°C | |
| MFCD00006765 | |
| 3077 | |
| NBYLBWHHTUWMER-UHFFFAOYSA-N | |
| 2-methylquinolin-8-ol | |
| 13224 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 826-81-3 | |
| 159.188 | |
| NBYLBWHHTUWMER-UHFFFAOYSA-N | |
| 13224 | |
| CC1=NC2=C(C=CC=C2O)C=C1 |
| C10H9NO | |
| MFCD00006765 | |
| 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol | |
| 2-methylquinolin-8-ol |
Safety and Handling
EINECSNumber : (5)-0808
RTECSNumber : VC7920000
TSCA : Yes