missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-8-quinolinol 98.0+%, TCI America™

Catalog No. p-7132160
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M042025G 25 g
M0420100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M042025G Supplier TCI America Supplier No. M042025G

Chemical Identifiers

CAS 826-81-3
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00006765
InChI Key NBYLBWHHTUWMER-UHFFFAOYSA-N
Synonym 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol
PubChem CID 13224
IUPAC Name 2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=CC=C2O)C=C1

Specifications

Melting Point 73°C
Boiling Point 145°C
Quantity 25 g
UN Number 3077
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-8-quinolinol
EINECSNumber (5)-0808
RTECSNumber VC7920000
TSCA Yes