missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-8-nitroquinoline, 98%

CAS: 881-07-2 | C10H8N2O2 | 188.19 g/mol

$26.48 - $79.27

Chemical Identifiers

CAS 881-07-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD00006764
InChI Key UHPGVDHXHDPYQP-UHFFFAOYSA-N
PubChem CID 13433
IUPAC Name 2-methyl-8-nitroquinoline
SMILES CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0300603
View Documents
Thermo Scientific Chemicals
L0300603
1 g
Each for $26.48
Only null left
 
AAL0300606
View Documents
Thermo Scientific Chemicals
L0300606
5 g
Each for $79.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

881-07-2
188.19
UHPGVDHXHDPYQP-UHFFFAOYSA-N
2-methyl-8-nitroquinoline
C10H8N2O2
MFCD00006764
13433
CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O

Specifications

881-07-2
Odorless
MFCD00006764
145077
UHPGVDHXHDPYQP-UHFFFAOYSA-N
2-methyl-8-nitroquinoline
13433
98%
136°C to 139°C
C10H8N2O2
1 g
8-nitroquinaldine, quinaldine, 8-nitro, quinoline, 2-methyl-8-nitro, unii-xrv949auqz, xrv949auqz, acmc-209qr6, 2-methyl-8-nitro-quinoline, 5-20-07-00387 beilstein handbook reference, quinoline,2-methyl-8-nitro, uhpgvdhxhdpyqp-uhfffaoysa
CC1=CC=C2C=CC=C(C2=N1)[N+]([O-])=O
188.19
188.19
2-Methyl-8-nitroquinoline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341
Suspected of causing genetic defects.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

EINECSNumber : 212-919-6

RTECSNumber : UZ9830000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only