missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitrobenzoic Anhydride 98.0+%, TCI America™
$104.38 - $1170.62
Chemical Identifiers
| CAS | 434935-69-0 |
|---|---|
| Molecular Formula | C16H12N2O7 |
| Molecular Weight (g/mol) | 344.279 |
| MDL Number | MFCD06797117 |
| InChI Key | YEKPNMQQSPHKBP-UHFFFAOYSA-N |
| Synonym | 6-Nitro-o-toluic Anhydride, MNBA |
| PubChem CID | 10472648 |
| IUPAC Name | (2-methyl-6-nitrobenzoyl) 2-methyl-6-nitrobenzoate |
| SMILES | CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC(=O)C2=C(C=CC=C2[N+](=O)[O-])C |
Chemical Identifiers
| 434935-69-0 | |
| 344.279 | |
| YEKPNMQQSPHKBP-UHFFFAOYSA-N | |
| 10472648 | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC(=O)C2=C(C=CC=C2[N+](=O)[O-])C |
| C16H12N2O7 | |
| MFCD06797117 | |
| 6-Nitro-o-toluic Anhydride, MNBA | |
| (2-methyl-6-nitrobenzoyl) 2-methyl-6-nitrobenzoate |
Specifications
| 434935-69-0 | |
| C16H12N2O7 | |
| 1 g | |
| YEKPNMQQSPHKBP-UHFFFAOYSA-N | |
| (2-methyl-6-nitrobenzoyl) 2-methyl-6-nitrobenzoate | |
| 10472648 | |
| ≥98.0% (T) | |
| 2-Methyl-6-nitrobenzoic Anhydride |
| 177°C | |
| MFCD06797117 | |
| 6-Nitro-o-toluic Anhydride, MNBA | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)OC(=O)C2=C(C=CC=C2[N+](=O)[O-])C | |
| 344.279 | |
| 344.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No