missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitrobenzoic Anhydride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 177°C |
| Quantity | 1 g |
| Formula Weight | 344.28 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Methyl-6-nitrobenzoic Anhydride |
Safety and Handling
| TSCA | No |