missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7132489
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M100210G 10 g
M100225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M100210G Supplier TCI America Supplier No. M100210G
Only null left

Chemical Identifiers

CAS 13506-76-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007267
InChI Key CCXSGQZMYLXTOI-UHFFFAOYSA-N
Synonym 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid
PubChem CID 16097
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 156°C
Quantity 10 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitrobenzoic Acid