missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitrobenzoic acid, 99%, Thermo Scientific™
Identifiants chimiques
| CAS | 13506-76-8 |
|---|---|
| Molecular Formula | C8H7NO4 |
| Molecular Weight (g/mol) | 181.15 |
| InChI Key | CCXSGQZMYLXTOI-UHFFFAOYSA-N |
| Synonym | 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid |
| PubChem CID | 16097 |
| SMILES | CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
AC165980050
|
N/A | 5g |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
| 13506-76-8 | |
| 181.15 | |
| 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O |
| C8H7NO4 | |
| CCXSGQZMYLXTOI-UHFFFAOYSA-N | |
| 16097 |
Spécifications
| 13506-76-8 | |
| 97.5% min. (HPLC) | |
| C8H7NO4 | |
| 09,I,189 | |
| Solubility in water: <0.1g/100mL (20°C). | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O | |
| 16097 | |
| 99% |
| Authentic | |
| Glass bottle | |
| CH3C6H3(NO2)CO2H | |
| 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid | |
| CCXSGQZMYLXTOI-UHFFFAOYSA-N | |
| 181.15 | |
| 181.15 | |
| 2-Methyl-6-nitrobenzoic acid |
Sécurité et manipulation
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
EINECSNumber : 236-833-3
RUO – Research Use Only