missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitrobenzoic acid, 99%, Thermo Scientific™
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Linear Formula | CH3C6H3(NO2)CO2H |
| Quantity | 5g |
| Beilstein | 09,I,189 |
| Solubility Information | Solubility in water: <0.1g/100mL (20°C). |
| Formula Weight | 181.15 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Methyl-6-nitrobenzoic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 236-833-3 |
RUO – Research Use Only