missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitrobenzoic acid, 98%

Catalog No. AAL0656314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0656314 25 g
AAL0656306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0656314 Supplier Thermo Scientific Chemicals Supplier No. L0656314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13506-76-8 | C8H7NO4 | 181.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13506-76-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007267
InChI Key CCXSGQZMYLXTOI-UHFFFAOYSA-N
Synonym 2-methyl-6-nitrobenzoic acid, 6-nitro-o-toluic acid, benzoic acid, 2-methyl-6-nitro, 6-methyl-2-nitrobenzoic acid, 2-methyl-6-nitro-benzoic acid, o-toluic acid, 6-nitro, 2-methyl-6-nitrobenzoicacid, unii-jm85y56dx1, nitrotoluylsaure, nitro-ortho-toluic acid
PubChem CID 16097
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 153°C to 156°C
Quantity 25 g
Beilstein 2050804
Formula Weight 181.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-833-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only