missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-6-nitroaniline 99.0+%, TCI America™
$240.46 - $848.14
Chemical Identifiers
| CAS | 570-24-1 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.153 |
| MDL Number | MFCD00007744 |
| InChI Key | FCMRHMPITHLLLA-UHFFFAOYSA-N |
| Synonym | 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc |
| PubChem CID | 11298 |
| IUPAC Name | 2-methyl-6-nitroaniline |
| SMILES | CC1=C(C(=CC=C1)[N+](=O)[O-])N |
Chemical Identifiers
| 570-24-1 | |
| 152.153 | |
| FCMRHMPITHLLLA-UHFFFAOYSA-N | |
| 11298 | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])N |
| C7H8N2O2 | |
| MFCD00007744 | |
| 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc | |
| 2-methyl-6-nitroaniline |
Specifications
| 570-24-1 | |
| 124°C | |
| MFCD00007744 | |
| 2660 | |
| FCMRHMPITHLLLA-UHFFFAOYSA-N | |
| 2-methyl-6-nitroaniline | |
| 11298 | |
| ≥99.0% (GC) | |
| 2-Methyl-6-nitroaniline |
| 96°C | |
| C7H8N2O2 | |
| 25 g | |
| 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc | |
| CC1=C(C(=CC=C1)[N+](=O)[O-])N | |
| 152.153 | |
| 152.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No