missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroaniline 99.0+%, TCI America™

Catalog No. N046725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N046725G 25 g
N0467250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N046725G Supplier TCI America Supplier No. N046725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 570-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007744
InChI Key FCMRHMPITHLLLA-UHFFFAOYSA-N
Synonym 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc
PubChem CID 11298
IUPAC Name 2-methyl-6-nitroaniline
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])N

Specifications

Melting Point 96°C
Boiling Point 124°C
Quantity 25 g
UN Number 2660
Formula Weight 152.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitroaniline