missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-6-nitroaniline, 98%

Catalog No. AAL0180322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0180322 100 g
AAL0180314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0180322 Supplier Thermo Scientific Chemicals Supplier No. L0180322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 570-24-1 | C7H8N2O2 | 152.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 570-24-1
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007744
InChI Key FCMRHMPITHLLLA-UHFFFAOYSA-N
Synonym 6-nitro-o-toluidine, 2-amino-3-nitrotoluene, benzenamine, 2-methyl-6-nitro, 2-methyl-6-nitrobenzenamine, 6-methyl-2-nitroaniline, 1-amino-2-methyl-6-nitrobenzene, unii-oup165ykbc, 3-nitro-2-aminotoluene, 2-nitro-6-methylaniline, oup165ykbc
PubChem CID 11298
IUPAC Name 2-methyl-6-nitroaniline
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])N

Specifications

Melting Point 93°C to 96°C
Quantity 100 g
UN Number UN2660
Beilstein 1868029
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-6-nitroaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROTOLUIDINES (MONO)
EINECSNumber 209-329-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only