Learn More
2-Methyl-5-phenyl-3-furoic acid, 97%
CAS: 108124-17-0 | C12H9O3 | 201.20 g/mol
Supplier: Thermo Scientific Chemicals B2098803
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methyl-5-phenyl-3-furoic acid | |
| 181°C to 184°C | |
| MFCD00221070 | |
| 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid | |
| CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O | |
| 201.20 | |
| 202.21 |
| 108124-17-0 | |
| C12H9O3 | |
| 1 g | |
| VLMNACSEESRUAK-UHFFFAOYSA-M | |
| 2-methyl-5-phenylfuran-3-carboxylic acid | |
| 736516 | |
| 97% |
Chemical Identifiers
| 108124-17-0 | |
| 201.20 | |
| VLMNACSEESRUAK-UHFFFAOYSA-M | |
| 736516 |
| C12H9O3 | |
| MFCD00221070 | |
| 2-methyl-5-phenyl-3-furoic acid, 3-furancarboxylic acid, 2-methyl-5-phenyl, 3-furancarboxylic acid,2-methyl-5-phenyl, acmc-1bptp, maybridge1_002369, cbmicro_037931, cambridge id 5930829, 2-methyl-5-phenyl-3-furancarboxylic acid, 2-methyl-5-phenyl-furan-3-carboxylic acid | |
| CC1=C(C=C(O1)C1=CC=CC=C1)C([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only