missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-oxo-1-cyclopentenyl Butyrate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 85°C |
| Quantity | 5 g |
| Formula Weight | 182.22 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methyl-5-oxo-1-cyclopentenyl Butyrate |
Safety and Handling
| EINECSNumber | (3)-2388 |
| TSCA | Yes |