accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 68227-51-0

IUPAC Name: 2-methyl-5-oxocyclopent-1-en-1-yl butanoate
Synonyms: 2-methyl-5-oxocyclopent-1-en-1-yl butanoate
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: SUJWBOKDKMEAOQ-UHFFFAOYSA-N
SMILES: CCCC(=O)OC1=C(C)CCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results