missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroimidazole-1-ethanol 99.0+%, TCI America™

Catalog No. p-7132445
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M092425G 25 g
M0924500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M092425G Supplier TCI America Supplier No. M092425G

Chemical Identifiers

CAS 443-48-1
Molecular Formula C6H9N3O3
Molecular Weight (g/mol) 171.156
MDL Number MFCD00009750
InChI Key VAOCPAMSLUNLGC-UHFFFAOYSA-N
Synonym metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
PubChem CID 4173
ChEBI CHEBI:6909
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
SMILES CC1=NC=C(N1CCO)[N+](=O)[O-]

Specifications

Melting Point 161°C
Color Yellow
Quantity 25 g
Formula Weight 171.16
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitroimidazole-1-ethanol
EINECSNumber (9)-1130
RTECSNumber NI5600000
TSCA No