missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-nitrobenzotrifluoride 98.0+%, TCI America™
$44.28 - $144.39
Chemical Identifiers
| CAS | 89976-12-5 |
|---|---|
| Molecular Formula | C8H6F3NO2 |
| Molecular Weight (g/mol) | 205.136 |
| MDL Number | MFCD01631684 |
| InChI Key | SVQCVQCIZWSPPX-UHFFFAOYSA-N |
| Synonym | 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 |
| PubChem CID | 2775447 |
| IUPAC Name | 1-methyl-4-nitro-2-(trifluoromethyl)benzene |
| SMILES | CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F |
Chemical Identifiers
| 89976-12-5 | |
| 205.136 | |
| SVQCVQCIZWSPPX-UHFFFAOYSA-N | |
| 2775447 | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F |
| C8H6F3NO2 | |
| MFCD01631684 | |
| 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 | |
| 1-methyl-4-nitro-2-(trifluoromethyl)benzene |
Specifications
| 89976-12-5 | |
| Yellow | |
| MFCD01631684 | |
| 2811 | |
| SVQCVQCIZWSPPX-UHFFFAOYSA-N | |
| 1-methyl-4-nitro-2-(trifluoromethyl)benzene | |
| 2775447 | |
| ≥98.0% (GC) | |
| 2-Methyl-5-nitrobenzotrifluoride |
| 39°C | |
| C8H6F3NO2 | |
| 1 g | |
| 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444 | |
| CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F | |
| 205.136 | |
| 205.14 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator