missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. M27981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27981G 1 g
M27985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27981G Supplier TCI America Supplier No. M27981G

Chemical Identifiers

CAS 89976-12-5
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.136
MDL Number MFCD01631684
InChI Key SVQCVQCIZWSPPX-UHFFFAOYSA-N
Synonym 2-methyl-5-nitrobenzotrifluoride, 1-methyl-4-nitro-2-trifluoromethyl benzene, 2-trifluoromethyl-4-nitrotoluene, 4-nitro-2-trifluoromethyl toluene, 1-methyl-4-nitro-2-trifluoromethyl-benzene, benzene, 1-methyl-4-nitro-2-trifluoromethyl, 3-trifluoromethyl-4-methylnitrobenzene, 2-trifluoromethyl-1-methyl-4-nitrobenzene, pubchem4444
PubChem CID 2775447
IUPAC Name 1-methyl-4-nitro-2-(trifluoromethyl)benzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 39°C
Color Yellow
Quantity 1 g
UN Number 2811
Formula Weight 205.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methyl-5-nitrobenzotrifluoride
TSCA No
Recommended Storage Refrigerator