missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroaniline 98.0+%, TCI America™

$95.06 - $1129.62

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=CC=C(C=C1N)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N011525G
View Documents
TCI America
N011525G
25 g
Each for $95.06
Only null left
 
N0115500G
View Documents
TCI America
N0115500G
500 g
Each for $1,129.62
Only null left
 
Description

Description

Specifications

Chemical Identifiers

99-55-8
152.15
DSBIJCMXAIKKKI-UHFFFAOYSA-N
7444
2-methyl-5-nitroaniline
C7H8N2O2
MFCD00007741
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
CHEBI:66891
CC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

99-55-8
C7H8N2O2
25 g
2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
CC1=CC=C(C=C1N)[N+]([O-])=O
152.15
CHEBI:66891
≥98.0% (GC,T)
2-Methyl-5-nitroaniline
109°C
MFCD00007741
2660
DSBIJCMXAIKKKI-UHFFFAOYSA-N
2-methyl-5-nitroaniline
7444
152.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0401&(5)-2306

RTECSNumber : XU8225000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'