missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroaniline 98.0+%, TCI America™

Catalog No. N011525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N011525G 25 g
N0115500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N011525G Supplier TCI America Supplier No. N011525G

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

Melting Point 109°C
Quantity 25 g
UN Number 2660
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitroaniline
EINECSNumber (3)-0401&(5)-2306
RTECSNumber XU8225000
TSCA Yes