missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(methylthio)furan, 99%

Catalog No. AAB2199804
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2199804 2 g
AAB2199809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2199804 Supplier Thermo Scientific Chemicals Supplier No. B2199804
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13678-59-6 | C6H8OS | 128.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13678-59-6
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.189
MDL Number MFCD01208018
InChI Key RESBOJMQOGJOMW-UHFFFAOYSA-N
Synonym 2-methyl-5-methylthio furan, 5-methyl-2-methylthio furan, furan, 2-methyl-5-methylthio, methyl 5-methylfuryl sulfide, 2-methyl-5-methylsulfanyl furan, 2-methyl-5-methylthiofuran, unii-41z7b950jj, 2-methyl 5-methyl thio furan, furan,2-methyl-5-methylthio, fema no. 3366
PubChem CID 61657
IUPAC Name 2-methyl-5-methylsulfanylfuran
SMILES CC1=CC=C(O1)SC

Specifications

Density 1.063
Boiling Point 79°C to 81°C (50 mmHg)
Flash Point 50°C (122°F)
Odor Stench
Quantity 2 g
UN Number UN1993
Formula Weight 128.19
Percent Purity 99%
Chemical Name or Material 2-Methyl-5-(methylthio)furan
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 237-170-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only