missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-isopropylaniline 93.0+%, TCI America™

Catalog No. M08491G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08491G 1 g
M08495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08491G Supplier TCI America Supplier No. M08491G

Chemical Identifiers

CAS 2051-53-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00044503
InChI Key YKWALWNGEXPARQ-UHFFFAOYSA-N
Synonym 2-Amino-p-cymene, 5-Isopropyl-2-methylaniline, 5-Isopropyl-o-toluidine
PubChem CID 137414
IUPAC Name 2-methyl-5-(propan-2-yl)aniline
SMILES CC(C)C1=CC=C(C)C(N)=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 149.24
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-isopropylaniline