accessibility menu, dialog, popup

UserName

CAS RN 2051-53-8

IUPAC Name: 2-methyl-5-(propan-2-yl)aniline
Synonyms: 5-isopropyl-2-methylaniline
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: YKWALWNGEXPARQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C(N)=C1
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)