missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-undecanone 97.0+%, TCI America™
$156.59 - $935.76
Chemical Identifiers
| CAS | 19594-40-2 |
|---|---|
| Molecular Formula | C12H24O |
| Molecular Weight (g/mol) | 184.323 |
| MDL Number | MFCD00027319 |
| InChI Key | IEBAJFDSHJYDCK-UHFFFAOYSA-N |
| Synonym | Heptyl Isobutyl Ketone |
| PubChem CID | 246924 |
| IUPAC Name | 2-methylundecan-4-one |
| SMILES | CCCCCCCC(=O)CC(C)C |
Chemical Identifiers
| 19594-40-2 | |
| 184.323 | |
| IEBAJFDSHJYDCK-UHFFFAOYSA-N | |
| 246924 | |
| CCCCCCCC(=O)CC(C)C |
| C12H24O | |
| MFCD00027319 | |
| Heptyl Isobutyl Ketone | |
| 2-methylundecan-4-one |
Specifications
| 19594-40-2 | |
| 124°C | |
| MFCD00027319 | |
| Heptyl Isobutyl Ketone | |
| CCCCCCCC(=O)CC(C)C | |
| 184.323 | |
| 184.32 | |
| Liquid |
| Yellow | |
| C12H24O | |
| 1 g | |
| IEBAJFDSHJYDCK-UHFFFAOYSA-N | |
| 2-methylundecan-4-one | |
| 246924 | |
| ≥97.0% (GC) | |
| 2-Methyl-4-undecanone |
Safety and Handling
EINECSNumber : (2)-2475
TSCA : Yes