missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(trifluoromethoxy)aniline, 97+%

Catalog No. p-7027162
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2370603 1 g
AAB2370606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2370603 Supplier Thermo Scientific Chemicals Supplier No. B2370603
Only null left

CAS: 86256-59-9 | C8H8F3NO | 191.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86256-59-9
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD01631541
InChI Key IIDBMILLZRYZCH-UHFFFAOYSA-N
Synonym 2-methyl-4-trifluoromethoxy aniline, 2-methyl-4-trifluoromethoxy benzenamine, 4-trifluoromethoxy-2-methylaniline, 4-trifluoromethoxy-o-toluidine, benzenamine, 2-methyl-4-trifluoromethoxy, 2-methyl-4-trifluoromethoxy phenylamine, pubchem8512, ksc495s8h, 2-amino-5-trifluoromethoxy toluene
PubChem CID 2775546
IUPAC Name 2-methyl-4-(trifluoromethoxy)aniline
SMILES CC1=C(C=CC(=C1)OC(F)(F)F)N

Specifications

Quantity 1 g
UN Number UN2810
Formula Weight 191.15
Percent Purity ≥97%
Chemical Name or Material 2-Methyl-4-(trifluoromethoxy)aniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only