missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-quinolinol 98.0+%, TCI America™

Catalog No. M26205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26205G 5 g
M262025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26205G Supplier TCI America Supplier No. M26205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 607-67-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00006758,MFCD00518775
InChI Key NWINIEGDLHHNLH-UHFFFAOYSA-N
Synonym 2-methylquinolin-4-ol, 4-hydroxy-2-methylquinoline, 2-methyl-4-quinolinol, 4-quinolinol, 2-methyl, 4-hydroxyquinaldine, 2-methyl-4-hydroxyquinoline, 2-methyl-quinolin-4-ol, 2-methylquinolin-4 1h-one, 2-methyl-4 1h-quinolinone, 2-methyl-1,4-dihydroquinolin-4-one
PubChem CID 69089
IUPAC Name 2-methyl-1,4-dihydroquinolin-4-one
SMILES CC1=CC(=O)C2=CC=CC=C2N1

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 5 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-quinolinol
EINECSNumber (5)-3741
TSCA No