missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-quinolinol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 235°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 159.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Methyl-4-quinolinol |
Safety and Handling
| EINECSNumber | (5)-3741 |
| TSCA | No |