missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitropyridine N-Oxide 98.0+%, TCI America™

Catalog No. p-7133032
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21145G 5 g
M211425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21145G Supplier TCI America Supplier No. M21145G
Only null left

Chemical Identifiers

CAS 5470-66-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00014625
InChI Key FTTIAVRPJGCXAC-UHFFFAOYSA-N
Synonym 4-nitro-2-picoline n-oxide, 2-methyl-4-nitropyridine 1-oxide, 2-methyl-4-nitropyridine n-oxide, 2-methyl-4-nitropyridine-n-oxide, pyridine, 2-methyl-4-nitro-, 1-oxide, 4-nitro-2-picoline-n-oxide, 4-nitropyridine-2-picoline-n-oxide, 2-methyl-4-nitropyridin-1-ium-1-olate, 2-methyl-4-nitropyridinium-1-olate, 4-nitro-2-methylpyridine-n-oxide
PubChem CID 95291
IUPAC Name 2-methyl-4-nitropyridin-1-ium-1-olate
SMILES CC1=CC(=CC=[N+]1[O-])[N+]([O-])=O

Specifications

Melting Point 156°C
Color Yellow
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-nitropyridine N-Oxide
RTECSNumber UT5710000
TSCA No