CAS RN 5470-66-6
IUPAC Name:
2-methyl-4-nitropyridin-1-ium-1-olate
Synonyms:
2-methyl-4-nitropyridin-1-ium-1-olate
Molecular Weight (g/mol):
154.13
Molecular Formula:
C6H6N2O3
InChi Key:
FTTIAVRPJGCXAC-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=[N+]1[O-])[N+]([O-])=O
1
–
1
of
1
results
1
2-Methyl-4-nitropyridine N-Oxide 98.0+%, TCI America™
CAS: 5470-66-6 Molecular Formula: C6H6N2O3 Molecular Weight (g/mol): 154.13 MDL Number: MFCD00014625 InChI Key: FTTIAVRPJGCXAC-UHFFFAOYSA-N Synonym: 4-nitro-2-picoline n-oxide,2-methyl-4-nitropyridine 1-oxide,2-methyl-4-nitropyridine n-oxide,2-methyl-4-nitropyridine-n-oxide,pyridine, 2-methyl-4-nitro-, 1-oxide,4-nitro-2-picoline-n-oxide,4-nitropyridine-2-picoline-n-oxide,2-methyl-4-nitropyridin-1-ium-1-olate,2-methyl-4-nitropyridinium-1-olate,4-nitro-2-methylpyridine-n-oxide PubChem CID: 95291 IUPAC Name: 2-methyl-4-nitropyridin-1-ium-1-olate SMILES: CC1=CC(=CC=[N+]1[O-])[N+]([O-])=O