missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-4-nitrobenzonitrile 98.0+%, TCI America™
$186.74 - $564.08
Chemical Identifiers
| CAS | 89001-53-6 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.148 |
| MDL Number | MFCD03095378 |
| InChI Key | RNTFKDBRMXYEPR-UHFFFAOYSA-N |
| Synonym | 4-Nitro-o-tolunitrile |
| PubChem CID | 4346367 |
| IUPAC Name | 2-methyl-4-nitrobenzonitrile |
| SMILES | CC1=C(C=CC(=C1)[N+](=O)[O-])C#N |
Chemical Identifiers
| 89001-53-6 | |
| 162.148 | |
| RNTFKDBRMXYEPR-UHFFFAOYSA-N | |
| 4346367 | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])C#N |
| C8H6N2O2 | |
| MFCD03095378 | |
| 4-Nitro-o-tolunitrile | |
| 2-methyl-4-nitrobenzonitrile |
Specifications
| 89001-53-6 | |
| C8H6N2O2 | |
| 1 g | |
| RNTFKDBRMXYEPR-UHFFFAOYSA-N | |
| 2-methyl-4-nitrobenzonitrile | |
| 4346367 | |
| ≥98.0% (GC) | |
| 2-Methyl-4-nitrobenzonitrile |
| 100°C | |
| MFCD03095378 | |
| 4-Nitro-o-tolunitrile | |
| CC1=C(C=CC(=C1)[N+](=O)[O-])C#N | |
| 162.148 | |
| 162.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No