missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitrobenzonitrile 98.0+%, TCI America™

Catalog No. M29651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29651G 1 g
M29655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29651G Supplier TCI America Supplier No. M29651G
Only null left

Chemical Identifiers

CAS 89001-53-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD03095378
InChI Key RNTFKDBRMXYEPR-UHFFFAOYSA-N
Synonym 4-Nitro-o-tolunitrile
PubChem CID 4346367
IUPAC Name 2-methyl-4-nitrobenzonitrile
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])C#N

Specifications

Melting Point 100°C
Quantity 1 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-nitrobenzonitrile