missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-nitroaniline 99.0+%, TCI America™

Catalog No. N052925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N052925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N052925G Supplier TCI America Supplier No. N052925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-52-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007734
InChI Key XTTIQGSLJBWVIV-UHFFFAOYSA-N
Synonym 2-amino-5-nitrotoluene, 4-nitro-o-toluidine, 5-nitro-2-aminotoluene, p-nitro-o-toluidine, benzenamine, 2-methyl-4-nitro, fast red rl base, 1-amino-2-methyl-4-nitrobenzene, diabase red rl, ansibases red rl, devol red rl
PubChem CID 7441
IUPAC Name 2-methyl-4-nitroaniline
SMILES CC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

Melting Point 132°C
Quantity 25 g
UN Number 2660
Formula Weight 152.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-nitroaniline
EINECSNumber (3)-0401
RTECSNumber XU8210000
TSCA Yes