missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4,5-diphenyloxazole, TCI America™

Catalog No. M12995G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12995G Supplier TCI America Supplier No. M12995G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14224-99-8
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.29
MDL Number MFCD00005307
InChI Key QLQIWRCWPJRJJA-UHFFFAOYSA-N
PubChem CID 139703
IUPAC Name 2-methyl-4,5-diphenyl-1,3-oxazole
SMILES CC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 212°C
Quantity 5 g
Formula Weight 235.29
Physical Form Liquid
Chemical Name or Material 2-Methyl-4,5-diphenyloxazole
EINECSNumber (5)-0183
TSCA No