accessibility menu, dialog, popup

UserName

CH3ON

CAS RN 14224-99-8

IUPAC Name: 2-methyl-4,5-diphenyl-1,3-oxazole
Synonyms: 2-methyl-4,5-diphenyl-1,3-oxazole
Molecular Weight (g/mol): 235.29
Molecular Formula: C16H13NO
InChi Key: QLQIWRCWPJRJJA-UHFFFAOYSA-N
SMILES: CC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results