missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-propylpyrazine 95.0+%, TCI America™

Catalog No. p-7132396
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08415ML 5 mL
M084125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08415ML Supplier TCI America Supplier No. M08415ML
Only null left

Chemical Identifiers

CAS 15986-80-8
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00010096
InChI Key XAWKNALRUSOTOY-UHFFFAOYSA-N
Synonym 2-methyl-3-n-propylpyrazine, pyrazine, 2-methyl-3-propyl, 2-propyl-3-methylpyrazine, 3-propyl-2-methylpyrazine, 2-methyl-3-propyl-pyrazine, 3-propyl-2-methyl pyrazine, 2-methyl-3-propylpyrazin, methylpropylpyrazine, acmc-209dk8, chembl95176
PubChem CID 85224
IUPAC Name 2-methyl-3-propylpyrazine
SMILES CCCC1=NC=CN=C1C

Specifications

Color Yellow
Boiling Point 190°C
Quantity 5 mL
Formula Weight 136.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-propylpyrazine